1,2,3-Trifluorbenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL324752-1GEA 43.7 EUR
SIAL324752-1G
1,2,3-Trifluorbenzol, Sigma-Aldrich®
1,2,3-Trifluorobenzene
Formel: C₆H₃F₃
Molecular Weight: 132,09 g/mol
Siedepunkt: 95 °C (1013 hPa)
Dichte: 1,28 g/cm³ (20 °C)
MDL: MFCD00012013
CAS-Nummer: 1489-53-8
UN: 2387
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR