1,2,3,4-Tetrafluorbenzol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIALT11630-5GEA
63.3
EUR
SIALT11630-5G
1,2,3,4-Tetrafluorbenzol, Sigma-Aldrich®
1,2,3,4-Tetrafluorobenzene
Formel:
C₆H₂F₄ Molecular Weight: 150,08 g/mol Siedepunkt: 95 °C (1013 hPa) Schmelzpunkt: –42 °C Dichte: 1,811 g/cm³ (20 °C) |
MDL:
MFCD00000285 CAS-Nummer: 551-62-2 EINECS: 208-997-6 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...