2,5-Dimethoxybenzoesäurechlorid

Lieferant: FLUOROCHEM LTD (CHEMPUR)

CHMPCFF167.2.5EA 268 EUR
CHMPCFF167.2.5 CHMPCFF167.10 FLUO014693-1G
2,5-Dimethoxybenzoesäurechlorid
2,5-Dimethoxybenzoesäurechlorid
Formel: C₉H₉ClO₃
Molecular Weight: 200,62 g/mol
CAS-Nummer: 17918-14-8
EINECS: 241-863-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR