2,5-Dichlorphenylisothiocyanat, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ quality level = MQ100

SIAL474932-5MLEA 73.5 EUR
SIAL474932-5ML
2,5-Dichlorphenylisothiocyanat, Sigma-Aldrich®
2,5-Dichlorphenylisothiocyanat
Formel: C₇H₃Cl₂NS
Molecular Weight: 204,08 g/mol
Siedepunkt: 145…147 °C (15 mmHg)
Dichte: 1,425 g/cm³ (20 °C)
MDL: MFCD00041054
CAS-Nummer: 3386-42-3
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR