3-Chlor-2-fluorbenzoylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Chlor-2-fluorbenzol-1-carbonylchlorid
M-Clarity™ quality level = MQ200
SIAL539554-1GEA
0
EUR
SIAL539554-1G
3-Chlor-2-fluorbenzoylchlorid, Sigma-Aldrich®
3-Chlor-2-fluorbenzoylchlorid
Formel:
C₇H₃Cl₂FO Molecular Weight: 193 g/mol Siedepunkt: 202…203 °C (1013 hPa) Dichte: 1,451 g/cm³ (20 °C) |
MDL:
MFCD01631548 CAS-Nummer: 85345-76-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...