3,5-Dichlorsalicylsäure
Lieferant: Molekula
Synonyme:
3,5-Dichlor-2-hydroxybenzoesäure
MOLE18351091-100GEA
0
EUR
MOLE18351091-100G
3,5-Dichlorsalicylsäure
3,5-Dichlorsalicylsäure
Formel:
C₇H₄Cl₂O₃ Molecular Weight: 207,01 g/mol Siedepunkt: 319 °C (1013 hPa) |
MDL:
MFCD00002442 CAS-Nummer: 320-72-9 EINECS: 206-281-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...