5-Chlor-2-methoxybenzoesäure

Lieferant: Molekula

Synonyme: 5-Chlor-o-anissäure

MOLE52004574-500GEA 0 EUR
MOLE52004574-500G
5-Chlor-2-methoxybenzoesäure
5-Chlor-2-methoxybenzoesäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
MDL: MFCD00017546
CAS-Nummer: 3438-16-2
EINECS: 222-343-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR