4-Amino-5-chlor-2-methoxybenzoesäure
Lieferant: CHEMPUR
CHMPRH01348.250EA
0
EUR
CHMPRH01348.250
4-Amino-5-chlor-2-methoxybenzoesäure
4-Amino-5-chlor-2-methoxybenzoesäure
Formel:
C₈H₈ClNO₃ Molecular Weight: 201,61 g/mol Siedepunkt: 354 °C (1013 hPa) Schmelzpunkt: 213…215 °C |
MDL:
MFCD00038794 CAS-Nummer: 7206-70-4 EINECS: 230-582-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...