3,4-Dichlorbenzhydrazid
Lieferant: Thermo Scientific
Synonyme: 3,4-Dichlorbenzohydrazid3,4-Dichlorbenzoesäurehydrazid3,4-Dichlorbenzol-1-carbohydrazid
MAYBTL00776DAEA 0 EUR
MAYBTL00776DA MAYBTL00776EA
3,4-Dichlorbenzhydrazid
3,4-Dichlorbenzhydrazid
Formel: C₇H₆Cl₂N₂O
Molecular Weight: 205,04 g/mol
MDL: MFCD00017057
CAS-Nummer: 28036-91-1