2-Octin-1-ol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL630837-5GEA 55.9 EUR
SIAL630837-5G
2-Octin-1-ol, Sigma-Aldrich®
2-Octin-1-ol
Formel: C₈H₁₄O
Molecular Weight: 126,2 g/mol
MDL: MFCD00039542
CAS-Nummer: 20739-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR