6-Heptinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL442879-1GEA 51 EUR
SIAL442879-1G
6-Heptinsäure, Sigma-Aldrich®
6-Heptynoic acid
Formel: C₇H₁₀O₂
Molecular Weight: 126,16 g/mol
MDL: MFCD00239430
CAS-Nummer: 30964-00-2
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR