2-Fluorzimtsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL222712-5GEA 55.8 EUR
SIAL222712-5G
2-Fluorzimtsäure, Sigma-Aldrich®
2-Fluorzimtsäure
Formel: C₉H₇FO₂
Molecular Weight: 166,15
MDL: MFCD00004370
CAS-Nummer: 451-69-4
EINECS: 207-195-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR