2-Fluorzimtsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO207100250EA
130
EUR
ACRO207100250
2-Fluorzimtsäure 98%
2-Fluorzimtsäure
Formel:
C₉H₇FO₂ Molecular Weight: 166,15 Schmelzpunkt: 179…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004370 CAS-Nummer: 451-69-4 EINECS: 207-195-3 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 179 to 183 °C |
HPLC | ≥97.5 % |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...