2-Hydroxyzimtsäure

Lieferant: Molekula
Achtung

MOLE49871841-100GEA 0 EUR
MOLE49871841-100G
2-Hydroxyzimtsäure
2-Hydroxyzimtsäure
Formel: C₉H₈O₃
MDL: MFCD00004379
CAS-Nummer: 614-60-8
EINECS: 210-386-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR