2-Methoxyzimtsäure ≥97,0%, Sigma-Aldrich®

Lieferant: Merck
Warning

Synonyme: 3-(2-Methoxyphenyl)acrylic acid,2-Methoxycinnamic Acid

65400-10G-F
SIAL65400-10G-FEA 59.3 EUR
SIAL65400-10G-F
2-Methoxyzimtsäure ≥97,0%, Sigma-Aldrich®
2-Methoxyzimtsäure
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,18 g/mol
MDL: MFCD00064238
CAS-Nummer: 6099-03-2
EINECS: 228-047-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR