4-Chlor-3-nitrozimtsäure

Lieferant: MENAI (PART OF CHEMPUR)

MENAIP31.50EA 0 EUR
MENAIP31.50
4-Chlor-3-nitrozimtsäure
4-Chlor-3-nitrozimtsäure
Formel: C₉H₆ClNO₄
Molecular Weight: 227,61
MDL: MFCD00063311
CAS-Nummer: 20797-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR