1,9-Diaminononan, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Nonamethylendiamin
, 1,9-Nonandiamin
M-Clarity™ quality level = MQ100
SIAL187127-5GEA
224
EUR
SIAL187127-5G
1,9-Diaminononan, Sigma-Aldrich®
1,9-Diaminononan
Formel:
C₉H₂₂N₂ Molecular Weight: 158,29 g/mol Siedepunkt: 258 °C (1013 hPa) Schmelzpunkt: 35…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008251 CAS-Nummer: 646-24-2 EINECS: 211-470-3 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...