1,1,1,5,5,6,6,6-Octafluor-2,4-hexandion, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
SIAL429503-1GEA
111
EUR
SIAL429503-1G
1,1,1,5,5,6,6,6-Octafluor-2,4-hexandion, Sigma-Aldrich®
1,1,1,5,5,6,6,6-Octafluor-2,4-hexandion
Formel:
C₆H₂F₈O₂ Molecular Weight: 258,07 g/mol Siedepunkt: 85…87 °C (1013 hPa) Dichte: 1,53 g/cm³ (20 °C) |
CAS-Nummer:
20825-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...