Fenofibrat ≥98% (durch HPLC)

Lieferant: Enzo Life Sciences
Achtung

ALEXALX270481G005EA 72.5 EUR
ALEXALX270481G005
Fenofibrat ≥98% (durch HPLC)
Fenofibrate

PPARα activator.


Hypolipidemic compound. PPARα (peroxisome proliferator-activated receptor α) agonist (EC₅₀ = 18 µM for mouse and EC₅₀ = 30 µM for human PPARα). Also binds to PPARγ, but with at least 10-fold less affinity and is inactive at PPARδ (up to EC₅₀ = 100 µM).


Soluble in acetone, ether, DMSO or dimethyl formamide; slightly soluble in 100% ethanol or methanol; insoluble in water.

Formel: C₂₀H₂₁ClO₄
Molecular Weight: 360,83 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00133314
CAS-Nummer: 49562-28-9
EINECS: 256-376-3
https://de.vwr.com/stibo/web/std.lang.all/75/97/45347597.jpg

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR