(R)-(-)-2-Phenylbuttersäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL279870-1GEA 108 EUR
SIAL279870-1G
(R)-(-)-2-Phenylbuttersäure, Sigma-Aldrich®
(R)-(-)-2-Phenylbuttersäure
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2 g/mol
MDL: MFCD00063165
CAS-Nummer: 938-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR