DL-m-Tyrosin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-3-(3-hydroxyphenyl)propionsäure3-Hydroxyphenylalanin3-(m-Hydroxyphenyl)-DL-alanin

M-Clarity™ quality level = MQ200

SIALT3629-1GEA 224 EUR
SIALT3629-1G
DL-m-Tyrosin, Sigma-Aldrich®
DL-m-Tyrosin
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Siedepunkt: 351 °C (1013 hPa)
Schmelzpunkt: 283 °C
Dichte: 1,456 g/cm³ (20 °C)
MDL: MFCD00002597
CAS-Nummer: 775-06-4
EINECS: 212-270-9
Merck Index: 12,09971

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR