Neomycin trisulphate salt hydrate pflanzliche Zellkulturen getestet, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
SIALN3144-100GEA
510
EUR
SIALN3144-100G
Neomycin trisulphate salt hydrate pflanzliche Zellkulturen getestet, Sigma-Aldrich®
Neomycin trisulphate salt hydrate
Formel:
C₂₃H₄₆N₆O₁₃·₃H₂SO₄·xH₂O Molecular Weight: 908,88 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150829 CAS-Nummer: 1405-10-3 EINECS: 215-773-1 Merck Index: 12,06542 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...