Cadmium ≥99,999% (Metall-Basis), Drahtform, Puratronic®, Ø 0.3 mm (0.013 in)

Gefahr

41567.G6EA 250.64 EUR
41567.G6
Cadmium ≥99,999% (Metall-Basis), Drahtform, Puratronic®, Ø 0.3 mm (0.013 in)
Cadmium
Formel: Cd
Molecular Weight: 112,41 g/mol
Siedepunkt: 765 °C (1013 hPa)
Schmelzpunkt: 321 °C
Dichte: 8,642 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010914
CAS-Nummer: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR