Mitochondria Isolation Kit
Lieferant: PromoKine
POMOPK-CA577-K288EA 369 EUR
POMOPK-CA577-K288
Mitochondria Isolation Kit
Protein-Extraktion Protein Extraction Kits
PromoKine's Mitochondria Isolation Kit (for tissue & cultured cells) offers two options for the isolation of intact mitochondria. The first option is utilising a reagent-based method allowing parallel processing of multiple samples. The second option uses traditional dounce homogenisation, which provides better mitochondrial yield.
Mitochondria are the powerhouse of the cells because they generate most of the supply of energy in the form of adenosine triphosphate (ATP). Mitochondria are double membrane organelles: an outer membrane and a folded inner membrane called cristae. Isolated mitochondria are the sample of choice to study mitochondrial respiration, assembly of the respiratory complexes, apoptosis, mtDNA and mtRNA, and mitochondrial protein profiling.