Mammalian Whole Cell Protein Extraction Kit
Lieferant: PromoKine
POMOPK-CA577-K269EA 255 EUR
POMOPK-CA577-K269
Mammalian Whole Cell Protein Extraction Kit
Protein-Extraktion Protein Extraction Kits
The Mammalian Whole Cell Protein Extraction Kit has been optimised for complete extraction of all cellular proteins.
  • Optimised lysis buffer and protease inhibitor cocktail included
  • Results in less than 20 minutes
The Mammalian Whole Cell Protein Extraction Kit contains an optimised lysis buffer and a protease inhibitor cocktail as well as other reagents for convenient extraction of mammalian proteins from tissues and cells under non denaturing conditions. The entire procedure takes less than 20 minutes, and the proteins can be used for many downstream applications such as gel electrophoresis, Western blotting, etc.