Mycoplasma-EX
Lieferant: PromoKine
POMOPK-CC91-4003EA 285 EUR
POMOPK-CC91-4003 POMOPK-CC91-4003-1
Mycoplasma-EX
Non-antibiotic/antibiotic solution for efficient elimination of mycoplasmas in cell culture
  • Can be used to clean most eukaryotic cell lines
  • Extremely low cytotoxicity: kills mycoplasmas very efficiently - but is safe for cells
  • Temperature stable, ready-to-use solution
  • No additional dosing during the treatment necessary
The most commonly used method for elimination, inactivation, or suppression of mycoplasmas in cell cultures is treatment with distinct antibiotics. In general, antibiotic therapies do not result in long-lasting, successful elimination. Also, the cytotoxic properties of antibiotics can cause undesirable side effects on eukaryotic cells and may facilitate the development of resistant mycoplasma strains.
The new Mycoplasma-EX method for mycoplasma elimination overcomes all of these drawbacks combining a non-antibiotic and a thoroughly adjusted antibiotic treatment resulting in unique features as compared to other methods and products. Mycoplasma-EX is the first biological reagent that quickly eliminates mycoplasmas by directly killing them, and not just by inhibiting their growth. It is the only anti-mycoplasma agent that can be used to clean most eukaryotic cell lines and primary cells as well as virus stocks fast and directly while showing an extremely low cytotoxicity.