Trihydroxyethylrutin ≥95%

Lieferant: Enzo Life Sciences

ALEXALX385030G025EA 173 EUR
ALEXALX385030G025
Trihydroxyethylrutin ≥95%
Trihydroxyethylrutin

More orally active than rutin. Vaso- and cardioprotective agent used clinically to treat venous disorders. Inhibits platelet aggregation. Protects biomembranes and DNA against the deleterious effects of γ-radiation. Free radical scavenger. Antioxidant.


  • Vaso- and cardioprotective agent


Light yellow to greenish powder.


Soluble in water, glycerol or propylene glycol.

Formel: C₃₃H₄₂O₁₉
Molecular Weight: 742,7 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00893813
CAS-Nummer: 7085-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR