Rufloxacin ≥98%

Lieferant: Enzo Life Sciences

ALEXALX380300M250EA 413 EUR
ALEXALX380300M250
Rufloxacin ≥98%
Rufloxacin

Fluoroquinolone antibacterial agent. Structurally similar to ofloxacin (Prod. No. ALX-380-297). Specific inhibitor of bacterial DNA gyrase (topoisomerase). Inhibits B-cell differentiation. Shows photosensitizing effects.

Formel: C₁₇H₁₈FN₃O₃S·HCl
Molecular Weight: 363,4 · 36,5 g/mol
CAS-Nummer: 106017-08-7
https://de.vwr.com/stibo/web/std.lang.all/05/63/45390563.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR