AMPIGENE® qPCR Probe Mix Lo-ROX
Lieferant: Enzo Life Sciences
ALEXENZNUC1051000EA 604 EUR
ALEXENZNUC1051000 ALEXENZNUC1050200
AMPIGENE® qPCR Probe Mix Lo-ROX
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzymes and Kits
Optimised probe-based qPCR.
AMPIGENE® qPCR Probe Mixes use the latest developments in polymerase technology and buffer chemistry to deliver high performing probe-based real-time PCR with minimal optimization. AMPIGENE® qPCR Probe Mixes are designed for use on a variety of probes including: Taqman ®, Molecular Beacons ®, and Scorpion probes ®. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE® qPCR Probe Mixes come with either Lo-ROX, Hi-ROX, or No-ROX concentration. For Hi-ROX and No-ROX, see Prod. Nos. ENZ-NUC106 and ENZ-NUC107. Choose the appropriate ROX concentration for the instrument (see instrument compatibility for reference dye chart below.)
Bestellinformation: Contains 2x AMPIGENE® qPCR Probe Mix Lo-ROX (includes dNTPs and polymerases).