Miltefosine 98%
Lieferant: Thermo Scientific
|
Gefahr
|
ACRO461830010EA
192
EUR
ACRO461830010
Miltefosine 98%
Miltefosine
Formel:
C₂₁H₄₆NO₄P Molecular Weight: 407,58 g/mol Schmelzpunkt: 216 °C (Zersetzung) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00133396 CAS-Nummer: 58066-85-6 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤7 % |
Titration with HClO4 | ≥97.5 % (on dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...