N-(4-Aminobutyl)-N-ethylisoluminol 90%

Lieferant: Thermo Scientific

ACRO439450010EA 478 EUR
ACRO439450010
N-(4-Aminobutyl)-N-ethylisoluminol 90%
N-(4-Aminobutyl)-N-ethylisoluminol
Formel: C₁₄H₂₀N₄O₂
Molecular Weight: 276,34 g/mol
Schmelzpunkt: 246…248,5 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00010559
CAS-Nummer: 66612-29-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥89.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR