Standards for water pollution QC check, complex nutrients, VHG
Lieferant: LGC Standards
PROOVHG-QWPCNUT-15EA 116 EUR
PROOVHG-QWPCNUT-15
Standards for water pollution QC check, complex nutrients, VHG
Standards Umweltanalyse-Standards
This solution is intended for use as a quality control check for methods related to complex nutrients.
This standard has been prepared as a concentrate and must be diluted prior to analysis. This concentrate is preserved with approximately 1% (v/v) hydrochloric acid.
Standards: Traceable to NIST SRM.