Turbidity calibration standards, polymer beads, Supelco®
Lieferant: Merck
TURB50P-500MLEA 0 EUR
TURB50P-500ML SUPLTURB100P-100ML TURB20P-100ML SUPLTURB2P-1L SUPLTURB2P-500ML SUPLTURB05P-500ML SUPLTURB500P-1L SUPLTURB10P-100ML SUPLTURB1P-100ML SUPLTURB2P-100ML SUPLTURB1P-1L SUPLTURB05P-100ML SUPLTURB2000P-100M SUPLTURB2000P-1L TURB1000P-1L SUPLTURB05P-1L SUPLTURB2000P-500M TURB20P-500ML SUPLTURB500P-100ML TURB10P-500ML SUPLTURB500P-500ML SUPLTURB100P-500ML
Turbidity calibration standards, polymer beads, Supelco®
Certified reference materials for turbidity measurement.
Format: Matrix material
Standards: Certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005.