Nitrite(N), ion chromatography standard, VHG
Lieferant: LGC Standards PROMOCHEM
VHG
PROOVHG-INO2N-500EA 100 EUR
PROOVHG-INO2N-500 PROOVHG-INO2N-100
Nitrite(N), ion chromatography standard, VHG
Standards Anionische Standards Nitrit (N)-Standards
This solution is intended for use as a certified reference material (CRM) calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Standards: Traceable to NIST and SRM.