Whole blood, human immunodeficiency virus (HIV)
HCMSPB000F-2-HIVEA 0 EUR
HCMSPB000F-2-HIV
Whole blood, human immunodeficiency virus (HIV)
Whole blood used for researching the causes, diagnosis, supportive care and treatment of the Human Immunodeficiency Virus (HIV). Collected from IRB-consented donors diagnosed with HIV or AIDS.
There is currently no cure for HIV or AIDS; however, with proper treatment we are able to keep the level of HIV virus in the body low. Patient samples can be used by researchers to investigate biomarker and diagnostic development, pre-clinical research validation, and drug toxicity and efficacy.
Storage: Use immediately upon receipt.
Bitte beachten Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.