Anti-CASP12 Rat Monoclonal Antibody [clone: 12G6]

Lieferant: ProSci
PRSI36-184EA 525 EUR
PRSI36-184
Anti-CASP12 Rat Monoclonal Antibody [clone: 12G6]
Antikörper
Caspase-12 is a member of the ICE/CED-3 family of caspases and is detected as a protein of ~51kDa. Overexpression of caspase-12 in cells induces apoptosis. Capase-12 is localized to the endoplasmatic reticulum (ER) and activated by ER stress and specifically cleaves and activates pro-caspase-9 and is also thought to be involved in the apoptosis induced by viral infection. Caspase-12-deficient mice are resistant to ER stress-induced apoptosis. Caspase-12 is probably expressed in many and perhaps all mouse tissues and is expressed at high levels in skeletal muscle, heart, lung and kidney and at moderate levels in brain and liver. In many humans caspase-12 is a pseudo gene and not expressed.

ELISA: (direct and indirect: 1:2,000-1:5,000). Immunoprecipitation: (1:500-1:5,000). Western Blot: (1:500-1:5,000 using ECL. Suggested blocking and dilution buffer is PBST containing 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: CASP12
Clonality: Monoclonal
Clone: 12G6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR