Anti-TNFRSF13B Rat Monoclonal Antibody (Biotin) [clone: 1A-10]
Lieferant: ProSci
PRSI36-146EA 612 EUR
PRSI36-146
Anti-TNFRSF13B Rat Monoclonal Antibody (Biotin) [clone: 1A-10]
Antikörper
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 1A-10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse