Anti-PCNA Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-463EA 655 EUR
PRSI49-463
Anti-PCNA Rabbit Polyclonal Antibody
Antikörper
The proliferating cell nuclear antigen (PCNA) is an auxiliary protein of DNA polymerase delta and is involved in the control of eukaryotic DNA replication by increasing the processibility of DNA polymerase during elongation of the leading strand. PCNA is expressed in the nucleus of all proliferating cells and is pivotal for DNA synthesis and cell cycle progression. In response to DNA damage, PCNA is mono-ubiquitinated and is involved in mismatch-provoked excision. PCNA is a useful marker for DNA synthesis and is highly conserved among most species.

PCNA antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 μg/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Fish, Xenopus, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR