Anti-CUL9 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-462EA 655 EUR
PRSI49-462
Anti-CUL9 Rabbit Polyclonal Antibody
Antikörper
Cullins assemble a potentially large number of ubiquitin ligases by binding to the RING protein ROC1 to catalyse polyubiquitination, as well as binding to various specificity factors to recruit substrates. PARC is a cullin family member that functions as a cytoplasmic anchor protein in p53-associated protein complexes. PARC regulates the subcellular localization of p53 and subsequent function. PARC forms a complex with p53 in the cytoplasm of unstressed cells and interacts with UBCH7 and UBCH8. PARC shows a cytoplasmic localization and is ubiquitously expressed in all tissues with highest expression in testis brain and kidney.

Cullin 9 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 μg/mL.

Delipidation and Defibrination

Sterile filtered antiserum, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CUL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR