Anti-ORM1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-461EA 619 EUR
PRSI49-461
Anti-ORM1 Rabbit Polyclonal Antibody
Antikörper
ORM1 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

IgG Fractionation

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ORM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR