Sitzung fortführen?
Automatische Abmeldung in: 
Sitzung abgelaufen!
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Anti-TNFRSF10A Mouse Monoclonal Antibody

Lieferant: ProSci Inc.
Cells undergo programmed cell death (Apoptosis) in response to various stimuli. Apoptosis is essential for morphogenesis, tissue homeostasis, and host defense Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel death domain containing receptor was recently identified and designated DR4 (for death receptor 4). The ligand for this novel death receptor has been identified and termed TRAIL (3,4), which is a new member in the TNF family. DR4 is also called TRAIL receptor-1 (TRAIL-R1). DR4 is expressed in most of human tissues including spleen, peripheral blood leukocytes, small intestine and thymus. Like TNFR1, Fas and DR3, DR4 mediates apoptosis and NF-κB activation in many tissues and cells. Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel death domain containing receptor was recently identified and designated DR4 (for death receptor 4)1. The ligand for this novel death receptor has been identified and termed TRAIL, which is a new member in the TNF family. DR4 is also called TRAIL receptor-1 (TRAIL-R1). DR4 is expressed in most of human tissues including spleen, peripheral blood leukocytes, small intestine and thymus. Like TNFR1, Fas and DR3, DR4 mediates apoptosis and NF-κB activation in many tissues and cells.

TNFRSF10A antibody can be used in immunohistochemistry starting at 10 μg/mL.

Protein G Column

PBS, 0.05% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

PRODUKT

Erweitert / Kompakt
Größe VE Katalognummer Menge
50 µg 50 µG PRSI49-324
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen

TECHNISCHE DATEN

Antigen symbol TNFRSF10A
Antigen name Tumor Necrosis Factor Receptor Superfamily, Member 10A
Reaktivität Human
Konjugation Unconjugated
Klonalität Monoclonal
Wirt Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Neutralisation Yes
Antigen synonyms DR4, CD261, TRAILR1, TRAILR-1, APO2
Immunogen TNFRSF10A monoclonal antibody was raised against a peptide corresponding to amino acid 1 to 20 of TNFRSF10A (Human).
Form Liquid
Tested applications Fluorescence-Activated Cell Sorting

BEWERTUNG VERFASSEN

Brauchen Sie Hilfe?

  • Was auch immer Sie zu sagen haben, positiv oder negativ, ist uns wichtig.
  • Das Bewerten dauert nur wenige Sekunden
  • Ihre Meinung wird öffentlich zur Verfügung gestellt.

VWR wird Ihren Namen als VWR Nutzer veröffentlichen.
Sie können diesen Namen nutzen oder vorher nochmal ändern und erneut abspeichern.
Wir empfehlen Initialien oder Spitznamen.

Bitte bestätigen Sie die Branche.

Beispiel: Pharma

Bitte bestätigen Sie Ihre Rolle.

Beispiel: Techniker

Bitte bestätigen Sie den Ort

Beispiel: Frankfurt

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

Weitere Angaben

Bitte wählen Sie den gewünschten Artikel

Bitte tragen Sie die Chargennummer ein. Sollte Ihnen diese nicht vorliegen, tragen Sie bitte bei "VWR-Chemikalien" ein "x" ein.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Kostengünstigere Alternativen prüfen & vergleichen
Wir haben eine kostengünstigere Alternative für Sie gefunden. Ihre Einsparung beträgt  pro Stück.  Sie können bis zu 3 Artikel miteinander vergleichen. Wählen Sie die gewünschten Artikel aus und drücken Sie auf Artikel vergleichen. Zum Ersetzen des ursprünglichen Artkels, geben Sie eine Menge im Alternativprodukt ein und klicken Sie auf Ersetzen.

Ursprünglicher Artikel:

Beschreibung Artikelnummer Verfügbarkeit Einheit Ihr Preis Preis pro Menge
 

Neues Design und unsere Verpflichtung unseren Kunden gegenüber

Verlassen Sie sich auf bekannte Funktionen und Ansprechpartner, kombiniert mit Innovation durch Avantor®. Unser neuer Markenauftritt symbolisiert Beweglichkeit und unsere Fähigkeit uns an die Anforderungen unserer Kunden anzupassen.

Learn More Start Shopping