Anti-SKIL Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-313EA 619 EUR
PRSI49-313
Anti-SKIL Rabbit Polyclonal Antibody
Antikörper
Principal Names: SnoN; SNO; SKI-like; Ski-related Oncogene; SnoA; SKIL Official Gene Symbol- SKIL Gen Bank Accession Number- NP_005405 Gene ID- 6498(Human) 20482 (mouse)Gene Map Locus- 3q26 (human) SnoN, an 80 kDa protein, is a member of ski family of nuclear proto-oncogenes involved in regulation of cellular transformation and differentiation. Primarily expressed in two isoforms, SnoN is localized in cytoplasm in normal tissues and non-tumorigenic primary epithelial cells. In cancer tissues or cells, SnoN is exclusively localized in the nucleus. It plays a vital role in inhibition of cell cycle arrest induced by TGF-. Upon morphological differentiation or cell-cycle arrest, SnoN translocates into the nucleus, binds to Smad2, Smad3, and Smad4 on TGF- -responsive promoters and represses their ability to activate expression of TGF- target genes. It has also been found to mediate transcriptional repression of thyroid hormone receptor, Mad and pRb. Increased expression of SnoN has been detected in many human tumor cell lines suggesting a clinical significance.

SKIL antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 2.5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SKIL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR