Anti-S100 A7 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-307EA 480 EUR
PRSI49-307
Anti-S100 A7 Mouse Monoclonal Antibody
Psoriasin/HID-5 was originally identified as an up-regulated protein in psoriatic keratinocytes. Subsequently, this protein was found to have elevated expression in abnormally differentiating primary keratinocytes and in various carcinomas, including ductal carcinoma in situ (2-3). Psoriasin/HID-5 may play a role in the establishment or maintenance of these pathologies, and may serve as a diagnostic marker of particular tumor types or abnormally proliferative epithelia.

S100A7 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS. Filter sterilized. Sodium azide free.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: S100 A7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR