Anti-OLFM4 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-306EA 619 EUR
PRSI49-306
Anti-OLFM4 Rabbit Polyclonal Antibody
Antikörper
Olfactomedin 4 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.05% BSA, 0.05% sodium azide.

PBS, 0.05% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OLFM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR