Anti-MAPK9 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-303EA 480 EUR
PRSI49-303
Anti-MAPK9 Rabbit Polyclonal Antibody
Antikörper
JNK2, a MAPK type protein kinase, regulates cell apoptosis, differentiation and growth in response to growth factors, inflammatory cytokines and cellular stress. JNK2 is phosphorylated by by MKK4/MAP2K4 and MKK7/MAP2K7. Activated JNK2 translocates to the nucleus where it phosphorylates transcription factors, including c-Jun, ATF2, ELK1, Jun-D and NFAT4. At least four distinct isoforms, resulting from alternatively spliced transcript variants, have been reported - three different isoforms comprised of 382 amino acids and one comprised of 424 amino acids.

MAPK9 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 1 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAPK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Bovine, Gibbon, Gorilla, Human, Pig, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR