Anti-LCK Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-299EA 480 EUR
PRSI49-299
Anti-LCK Mouse Monoclonal Antibody
Lck is a lymphoid-specific cytosolic protein tyrosine kinase (PTK), which is essential for T-cell development and function. It is constitutively associated with the cytoplasmic portions of the CD4 and CD8 surface receptors and plays a key role in T-cell antigen receptor (TCR)-linked signal transduction pathways. Association of the TCR with a peptide antigen-bound MHC complex facilitates the interaction of CD4 and CD8 with MHC class II and class I molecules, respectively, and thereby recruits the associated Lck to the vicinity of the TCR/CD3 complex. Lck then phosphorylates tyrosine residues within the immunoreceptor tyrosine-based activation motifs (ITAMs) in the cytoplasmic tails of the TCR-chains and CD3 subunits. The phospho-ITAMs serve as docking sites for Src homology domain 2 (SH2)-containing molecules, predominantly ZAP-70 and Syk. Only then can ZAP-70 undergo tyrosine phosphorylation, become enzymatically active and further phosphorylate downstream effector molecules. In addition, Lck contributes to signaling by other receptor molecules. Protein tyrosine kinase LCK is a non-receptor kinase belonging to the Src subfamily. LCK functions in a variety of ways, and therefore results in an assortment of diseases when not working properly. For example, T cell receptor binding activates LCK to induce downstream effects. In addition, LCK regulates chloride channel opening in lymphocytes. LCK is associated with various cancers and diseases, including neuroblastoma, Non-Hodgkins lymphoma, leukemia, SCID (severe combine immunodeficiency), and type I diabetes. Alternative splicing and differential promoter usage have been observed.

LCK antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 10 - 20 μg/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LCK
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR