Anti-IRAK2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-297EA 613 EUR
PRSI49-297
Anti-IRAK2 Rabbit Polyclonal Antibody
Antikörper
The binding of interleukin-1 (IL-1) to its receptor IL-1RI initiates a signaling cascade that results in the activation of NF-κB. Recently protein kinases designated IRAKs (IL-1 receptor-associated kinases) have been identified and designated as IRAK-1, IRAK-2 and IRAK-M. IRAK-2 is a serine/threonine protein kinase. The primary amino acid sequence of IRAK-2 shares similarity with that of Pelle, a protein kinase that is essential for the activation of a NF-κB homolog in Drosophila. IRAK (IL-1 receptor kinase) and mediates NF-κB activation by members of IL-1 family including IL-1 and a novel cytokine IL-18. Disruption of IRAK expression by anti-sense oligonucleotides inhibits IL-1 and IL-18-induced activation of NF-κB and activator protein, AP-1. IRAK2, an IRAK type protein kinase, is a signal transducer for the immune response interleukin-1 receptor (IL-1R) signaling cascade. IRAK2 also has been associated with signaling cell death, potentially via the Fas-associated death domain protein/caspase-8 apoptotic pathway. Dominant negative forms of IRAK2 attenuate IL-1R-mediated NF-kappa B activation, suggesting that IRAK-2 may serve as a therapeutic target for inhibiting IL-1-induced inflammation.

IRAK2 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IRAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR