Anti-CHD4 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-289EA 0 EUR
PRSI49-289
Anti-CHD4 Rabbit Polyclonal Antibody
The CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. Patients with dermatomyositis develop antibodies against the nuclear antigen chromodomain helicase DNA binding protein 4. The protein exists in a complex containing histone deacetylase and nucleosome remodeling activities, suggesting a role for chromatin reorganization in cancer metastasis.

CHD4 antibody can be used in ELISA starting at 1:2000 - 1:10000, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 1:500, and immunoprecipitation

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHD4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR