Anti-GAD1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-276EA 480 EUR
PRSI49-276
Anti-GAD1 Rabbit Polyclonal Antibody
Antikörper
GAD67, a 67 kDa protein, is a neuronal isoform of this enzyme predominantly expressed in human Brain. It consists of Pyridoxal 5-phosphate binding site and conserved Cysteine residues which are vital for its activity and is inhibited due to phosphorylation. It has high binding affinity for the cofactor Pyridoxal-5phosphate and its role has been implicated in normal Palate development. It is found to be associated with childhood onset schizophrenia and craniofacial malformations and has been identified as a major auto antigen and an auto-reactive T-cell target in IDDM. Deficiency of this enzyme leads to pyridoxine dependency along with epilepsy. It also might play a role in stiff main syndrome.

GAD1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR