Anti-EIF4G2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-274EA 480 EUR
PRSI49-274
Anti-EIF4G2 Mouse Monoclonal Antibody
Antikörper
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Recently, five novel apoptotic genes (named DAP genes for: Death Associated Proteins) have been identified that code for proteins, which display a diverse spectrum of biochemical activities. These include a novel type of calcium/calmodulin-regulated kinase, which carries ankyrin repeats and a death domain (DAP-kinase), a nucleotide-binding protein (DAP-3), a small proline-rich cytoplasmic protein (DAP-1), and a novel homolog of the eIF4G translation initiation factor (DAP-5). DAP-5 (also named p97 and NAT1) is a ubiquitously expressed member of the translation initiation factor eIF4G family that lacks the eIF4E binding site. DAP-5 was cloned from HeLa cells using a functional approach in attempt to isolate cDNA fragments, which convey resistance to gamma interferon (IFN-g)-induced programmed cell death. A fragment of the cDNA when expressed as a mini-protein of 28 kDa protects the cell from IFN-g induced apoptosis. The full-length DAP-5 cDNA codes for a protein of approximate molecular weight of 97 kDa. Low level of expression protects the cells, whereas high level of expression results in cell death (3-4).

EImmunofluorescence4G2 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EIF4G2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR